Learn More
Cayman Chemical Wilforlide A, Purity: ≥95%, CAS: 84104-71-2, 10 mg
SDP
Catalog No. 502949922
Encompass_Preferred
A triterpene; inhibits xylene-induced ear swelling in mice at 60 or 300 µg/kg; reduces inflammation, bone damage, and synovial hyperplasia in the inflamed joints in a rat model of collagen-induced arthritis in combination with triptolide
Catalog No. 50-294-9922 Supplier Cayman Chemical Supplier No. 3465410 MG
A triterpene; inhibits xylene-induced ear swelling in mice at 60 or 300 µg/kg; reduces inflammation, bone damage, and synovial hyperplasia in the inflamed joints in a rat model of collagen-induced arthritis in combination with triptolide