Learn More
TARGETMOL CHEMICALS INC Wilforlide A 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T338220MG
Encompass_Preferred
Wilforlide A (3-Epiabruslactone A) is a natural product, separated from the ethanolic extract of tripterygium wilfordii. High-dose wilforlide A has obvious anti-inflammatory effect, but has no significant immune suppressive activity. Purity ≥95%
Catalog No. 50-287-6844