Learn More
Apexbio Technology LLC Wilforlide A 84104-71-2 20mg
SDP
Catalog No. 5000094457
Encompass_Preferred
Wilforlide A (CAS nan) is a sesquiterpene lactone derivative isolated from Tripterygium wilfordii It is designed to modulate various signaling pathways involved in inflammatory responses immunological regulation and cellular proliferation processes thereby regulating immune and inflammatory pathways Wilforlide A exerts its biological activity primarily through the modulation of molecular mechanisms associated with inflammation-related disorders and immunomodulatory functions Based on these pharmacological properties Wilforlide A holds research potential in the investigation of inflammation immune regulation and potential antitumor activities
Catalog No. 50-000-94457 Supplier Apexbio Technology LLC Supplier No. N283220
Wilforlide A (CAS nan) is a sesquiterpene lactone derivative isolated from Tripterygium wilfordii It is designed to modulate various signaling pathways involved in inflammatory responses immunological regulation and cellular proliferation processes thereby regulating immune and inflammatory pathways Wilforlide A exerts its biological activity primarily through the modulation of molecular mechanisms associated with inflammation-related disorders and immunomodulatory functions Based on these pharmacological properties Wilforlide A holds research potential in the investigation of inflammation immune regulation and potential antitumor activities