Learn More
Bioworld Wilkins Chalgren Agar, 500 g
Catalog No. 50254418
Encompass_Preferred
Wilkins Chalgren AgarUsed to determin minimal inhibitory concentration (MIC’s) of antibiotics for anaerobic bacteria by the agar dilution procedurepH: 7.1Working Concentration: 48 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: yellow-beige agarSoluble in boiling waterContains:Enzymatic Digest of CaseinEnzymatic Digest of GelatinYeast ExtractSodium ChlorideDextroseL-ArginineSodium PyruvateHeminVitamin KAgar
Catalog No. 50-254-418 Supplier Bioworld Supplier No. 306201141
Wilkins Chalgren AgarUsed to determin minimal inhibitory concentration (MIC’s) of antibiotics for anaerobic bacteria by the agar dilution procedurepH: 7.1Working Concentration: 48 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: yellow-beige agarSoluble in boiling waterContains:Enzymatic Digest of CaseinEnzymatic Digest of GelatinYeast ExtractSodium ChlorideDextroseL-ArginineSodium PyruvateHeminVitamin KAgar