Learn More
Sigma Aldrich Fine Chemicals Biosciences Williams’ Medium E, 500mL
Catalog No. NC1684325
Encompass
With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture.
Catalog No. NC1684325 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W1878500ML
With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture.