Learn More
ELABSCIENCE William's E (without L-glutamine) 500mL
Catalog No. NC3523474
Encompass_Preferred
William's E medium was designed by William and Gunn. It is mainly used for the long-term culture of rat liver epithelial cells, and can also be used for the culture of other mammalian liver cells. L-glutamine is a necessary nutrient element in cell culture, but it is unstable in solution and will spontaneously degrade. The medium without L-glutamine can add fresh L-glutamine or its substitute arbitrarily according to research needs.William's medium contains many kinds of amino acids, vitamins, inorganic salts and other ingredients for cell culture, but does not contain protein, lipids or any growth factors. Therefore, the product should be used with serum or serum-free additives.
Catalog No. NC3523474 Supplier ELABSCIENCE Supplier No. PM151211
William's E medium was designed by William and Gunn. It is mainly used for the long-term culture of rat liver epithelial cells, and can also be used for the culture of other mammalian liver cells. L-glutamine is a necessary nutrient element in cell culture, but it is unstable in solution and will spontaneously degrade. The medium without L-glutamine can add fresh L-glutamine or its substitute arbitrarily according to research needs.William's medium contains many kinds of amino acids, vitamins, inorganic salts and other ingredients for cell culture, but does not contain protein, lipids or any growth factors. Therefore, the product should be used with serum or serum-free additives.