Learn More
Med Vet International Willoughby Tubing, Drip Chamber and Tubing Included, 20 drops/mL, J0064T, Each
Catalog No. 5000581460
Encompass_Preferred
Complete Infusion Set that can handle the large volume of fluid therapy necessary in large animals. The Willoughby style tubing stretches to 10', allowing movement to a restrained or stalled animal. It has a collapsable container made for easy storage in a mobile unit. Non-sterile.
Catalog No. 50-005-81460 Supplier Med Vet International Supplier No. JOR64T
Complete Infusion Set that can handle the large volume of fluid therapy necessary in large animals. The Willoughby style tubing stretches to 10', allowing movement to a restrained or stalled animal. It has a collapsable container made for easy storage in a mobile unit. Non-sterile.