Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM
Catalog No. p-6962025 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Type:
1,1800 Compound Database
3,500 Compound Database
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
16-800-207 1,1800 Compound Database
16-800-206 3,500 Compound Database
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-800-207 Supplier Bel-Art™ Supplier No. 660000107
Allows for Spectra processing using Zoom, Peak Integration and Peak Labeling. The Bel-Art SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM is an electronic book that contains high-resolution 300MHz 1H and 75MHz 13C spectra, integral, chemical structure and technical data for the more than 11,800 compounds.
  • Supplemental library that includes spectra for 3,500 additional compounds not included in the original printed version is also available
  • Order both the standard and the supplemental library for access to spectra on over 15,000 compounds
  • Multiple search methods: text Search by Chemical Name, Aldrich Catalog Number, CAS Number, Molecular Formula, advanced data field search on chemical categories and physical properties, such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight and Refractive Index
  • Minimum System Requirement: Windows Platform, CD-ROM drive, 600MB hard disk space
Specifications
Type 1,1800 Compound Database
Description FT-NMR Library with Spectral Viewer on CD-ROM