Learn More
Bel-Art™ Wilmad™ Kel-F Positioning Rod Microcell Small Volume NMR Insert
Shop All SP Bel-Art & SP Wilmad ProductsDescription
- Sample volume can be as little as 18 μl
- 3 different inserts with various shapes (Spherical Bulb, Capillary Tube and Cylindrical Bulb)
- Outer tube (Precision Thin Walled) must be ordered separately depending on magnetic field strength.
- Not recommended for variable temperature experiments.
- Use our PTFE Needles (#90630) with syringes to fill and clean sample.
Specifications
Specifications
| For Use With (Equipment) | 529-B adapter, Vortex Plugs WG-805C and under |
| Type | Positioning Rod |
| Description | Positioning Rod for inserting small volume microcells |
| Model | 529-C |
Only use with Wilmad-LabGlass Precision Thin-Walled Tubes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.