Description
- Low cost alternative to the full VT version for sample changers.
- Weight and susceptibility matched to originals.
- Can be mixed with originals in sample changer operation.
- VT operation of -150°C to 200°C
- Does not jam at top of upper barrel during insert operation.
- Tachometer strip now with foil protection for improved life span.
- Weight compatible with room temperature spinners.
- Very cost effective as compared to the originals.
Specifications
Specifications
| For Use With (Equipment) | Agilent/Varian NMR Spectrometer |
| Type | Spinner Turbine |
| Model | V-GFK-10 |