Learn More
Cambridge Isotope Laboratories Wilmad Quartz Non-Precision 4 Mm 250 Mm Length Epr Sample Tube, 1 Tube
Catalog No. 501629011
Encompass
Wilmad Quartz Non-Precision 4 Mm 250 Mm Length Epr Sample Tube, 1 Tube
Catalog No. 50-162-9011 Supplier Cambridge Isotope Laboratories Supplier No. 707SQ250M
Wilmad Quartz Non-Precision 4 Mm 250 Mm Length Epr Sample Tube, 1 Tube