Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR coaxial insert com1EA
Catalog No. 502901193
Encompass
The coaxial set is recommended for high field experiments under 600 MHz.featuresSwitch between three unique sample ratios during external referencing experimentsIdeal for variable temperature experiments as materials is the same between the tubes and spacerIn the complete set the insert and outer tube are fused together at the bottom
Catalog No. 50-290-1193 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5694451EA
The coaxial set is recommended for high field experiments under 600 MHz.featuresSwitch between three unique sample ratios during external referencing experimentsIdeal for variable temperature experiments as materials is the same between the tubes and spacerIn the complete set the insert and outer tube are fused together at the bottom