Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR NMR tubes 10 mm di
Catalog No. 502891525
Encompass
Made of Pyrex(R) borosilicate glass for compatibility with ground joints, valves, and vacuum racks. Precision ground and highly polished for spinning stability and highest resolution. Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes are thin-walled. Tubes that are 9 in. L are for making standards and for use with tip-off manifolds. Precision Wilmad NMR tubes are suitable for high field NMR spectroscopy.
Catalog No. 50-289-1525 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2747471PAK
Made of Pyrex(R) borosilicate glass for compatibility with ground joints, valves, and vacuum racks. Precision ground and highly polished for spinning stability and highest resolution. Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes are thin-walled. Tubes that are 9 in. L are for making standards and for use with tip-off manifolds. Precision Wilmad NMR tubes are suitable for high field NMR spectroscopy.