Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR NMR tubes 3 mm dia
Supplier: Sigma Aldrich Fine Chemicals Biosciences WIMWG300075050EA
With the steady increase of SNR on modern NMR spectrometers the trend of small volume testing fits in high-throughput NMR to save on solvent costs. The new 4 in. version is designed for Bruker(R) SampleJet(TM) system and possesses a tighter tolerance for use with the SampleJet(TM) cap which is sold separately.