Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR NMR tubes 3 mm dia
Catalog No. 502905971
Encompass
Made of Pyrex(R) borosilicate glass for compatibility with ground joints valves and vacuum racks. Precision ground and highly polished for spinning stability and highest reSOLUTIONon. Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes are thin-walled. Tubes that are 9 in. L are for making standards and for use with tip-off manifolds. These precision Wilmad NMR tubes are suitable for microscale NMR spectroscopy applications.
Catalog No. 50-290-5971 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2718615PAK
Made of Pyrex(R) borosilicate glass for compatibility with ground joints valves and vacuum racks. Precision ground and highly polished for spinning stability and highest reSOLUTIONon. Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes are thin-walled. Tubes that are 9 in. L are for making standards and for use with tip-off manifolds. These precision Wilmad NMR tubes are suitable for microscale NMR spectroscopy applications.