Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR NMR tubes 5 mm dia
Catalog No. 502901328
Encompass
Made of Pyrex(R) borosilicate glass (unless otherwise noted) for compatibility with ground joints valves and vacuum racks. Precision ground and highly polished for spinning stability and highest reSOLUTIONon.Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes are thin-walled unless otherwise stated. Tubes that are 9 in. L are for making standards and for use with tip-off manifolds. Precision Wilmad NMR tubes are suitable for high field NMR spectroscopy. Caps included.
Catalog No. 50-290-1328 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2745501PAK
Made of Pyrex(R) borosilicate glass (unless otherwise noted) for compatibility with ground joints valves and vacuum racks. Precision ground and highly polished for spinning stability and highest reSOLUTIONon.Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes are thin-walled unless otherwise stated. Tubes that are 9 in. L are for making standards and for use with tip-off manifolds. Precision Wilmad NMR tubes are suitable for high field NMR spectroscopy. Caps included.