Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR quartz CfQ EPR tub
Catalog No. 502901436
Encompass
Clear fused-Quartz (CfQ) is a naturally occurring material. It has a high dielectric constant and low levels of paramagnetic impurities. This NMR tube is suitable for use in routine testing in EPR spectropscopy excluding UV irradiation and samples with low sensitivity.
Catalog No. 50-290-1436 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5673965EA
Add to Cart
Add to Cart
Clear fused-Quartz (CfQ) is a naturally occurring material. It has a high dielectric constant and low levels of paramagnetic impurities. This NMR tube is suitable for use in routine testing in EPR spectropscopy excluding UV irradiation and samples with low sensitivity.