Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR quartz CfQ EPR tub
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z5665355EA
Clear fused-Quartz (CfQ) is a naturally occurring material. It has a high dielectric constant and low levels of paramagnetic impurities. This NMR tube is suitable for use in routine testing in EPR spectropscopy excluding UV irradiation and samples with low sensitivity.