Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR quick pressure val1EA
Catalog No. 502900742
Encompass
Specially designed to simplify the work of NMR scientists performing catalytic gas-liquid phase reactions air sensitive samples and elevated temperature studies using low boiling point solvents and can be used for EPR spectrometry too.The Quick Pressure Valve can also be attached to a quartz sample tube for EPR by using a graded seal that connects the Pyrex portion of the valve to the quartz tube.Easy to operate - one turn to open one turn to closeLarger opening for convenient sample additionLight weight concentric design for better performanceUnions and fittings for both 1/16 in. and 1/8 in. tubing
Catalog No. 50-290-0742 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5628661EA
Specially designed to simplify the work of NMR scientists performing catalytic gas-liquid phase reactions air sensitive samples and elevated temperature studies using low boiling point solvents and can be used for EPR spectrometry too.The Quick Pressure Valve can also be attached to a quartz sample tube for EPR by using a graded seal that connects the Pyrex portion of the valve to the quartz tube.Easy to operate - one turn to open one turn to closeLarger opening for convenient sample additionLight weight concentric design for better performanceUnions and fittings for both 1/16 in. and 1/8 in. tubing