Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR screw-cap NMR tube1EA
Catalog No. 502900488
Encompass
Precision Wilmad NMR tubes are suitable for high field NMR spectroscopy. NMR tube has an open-top screw-thread cap with a PTfE/silicone septum that permits addition of reactants or samples via syringe through the cap. Also good for sample storage. Caps and septa are replaceable.Excellent for handling air-sensitive samples and in cell perfusion studies.5 mm diameter tubes have size 8-425 caps (replacement Z162566)10 mm diameter tubes have size 15-425 caps (replacement Z106518)
Catalog No. 50-290-0488 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2719501EA
Precision Wilmad NMR tubes are suitable for high field NMR spectroscopy. NMR tube has an open-top screw-thread cap with a PTfE/silicone septum that permits addition of reactants or samples via syringe through the cap. Also good for sample storage. Caps and septa are replaceable.Excellent for handling air-sensitive samples and in cell perfusion studies.5 mm diameter tubes have size 8-425 caps (replacement Z162566)10 mm diameter tubes have size 15-425 caps (replacement Z106518)