Learn More
Sigma Aldrich Fine Chemicals Biosciences WilmadR Suprasil EPR tubes1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5674341EA
Encompass
Technological advances have increased the sensitivity in EPR spectroscopy causing certain high Q cavities to pick up the signal from the impurities in natural CfQ quartz tubes. When this is the case Suprasil EPR products which are made of synthetic material are a better option because they can supress major impurity contents. Tubes come with cap.
Catalog No. 50-290-0782