Learn More
World Precision Instrument Wilmer Iris Scissors, 10.5cm, Angled
SDP
Catalog No. 502539531
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-253-9531 Supplier World Precision Instrument Supplier No. 503248
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.