Learn More
Wimshurst Machine
SDPSCI_ED
Catalog No. s94229
Change view
Click to view available options
Wimshurst machine
Catalog No.
S94229 Wimshurst machine
1 options
Catalog No. S94229 Supplier Science First Supplier No. 6153190
Reproduces early electrical experiments.
  • Two high-resistance plastic discs have equally spaced metal sections
  • Discs are supported with two upright posts
  • Hand crank rotates discs in opposite directions
  • Produces substantial opposite charges which are deposited in the capacitors and on the metal electrodes
  • Collect induced charge with the brushes and adjust the electrodes and Leyden jar capacitors for higher potential
  • Generate sparks by lowering the system's capacitance
  • Mounted on an attractive wooden base
  • Disc dimensions: 25cm dia.
  • Instructions include theory and background information
Specifications
For Use With (Application) Demonstrates early electrical experiments with substantial opposite charges which are deposited in the capacitors and on the Metal electrodes
Includes 25cm dia. Two high resistance Plastic Acrylic discs with equally spaced Metal section, hand crank, Instructions