Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Wimshurst Machine
GREENER_CHOICESCI_ED

Catalog No. S139442 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139442 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139442 Supplier Supertek™ Supplier No. PH45120
Add to Cart
Add to Cart
This item is not returnable. View return policy

This Supertek™ Wimshurst Machine is a traditional electrostatic generator.

  • Teach a little history too, while using this traditional electrostatic generator.
  • History: The Wimshurst influence machine is an electrostatic generator, a machine for generating high voltages developed between 1880 and 1883 by British inventor James Wimshurst. It has a distinctive appearance with two large contra-rotating discs mounted in a vertical plane, two crossed bars with metallic brushes, and a spark gap formed by two metal spheres.
  • Supertek's electrostatic generator consists of two - 9.84 in. discs with radial aluminum segments driven in opposite directions by a hand wheel and belt with two Leyden jars on wooden base.

Specifications

Grade 6+
Format Model
Languages English
Subject Physical Science - Electricity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.