Learn More
Medchemexpress LLC HY-W011094 50mg Medchemexpress, Win 18446 CAS:1477-57-2 Purity:>98%
SDP
Catalog No. 502083308
Encompass_Preferred
Medchemexpress, HY-W011094 50mg Win 18446 CAS:1477-57-2 Win 18446 is an orally active testes-specific enzyme ALDH1a2 inhibitor, with an IC50 of 0.3 μM. Win 18446 reversibly inhibits spermatogenesis in many species and inhibits Retinoic acid (HY-14649) biosynthesis from Retinol (HY-B1342) within the testes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3308 Supplier Medchemexpress LLC Supplier No. HYW01109450MG
Medchemexpress, HY-W011094 50mg Win 18446 CAS:1477-57-2 Win 18446 is an orally active testes-specific enzyme ALDH1a2 inhibitor, with an IC50 of 0.3 μM. Win 18446 reversibly inhibits spermatogenesis in many species and inhibits Retinoic acid (HY-14649) biosynthesis from Retinol (HY-B1342) within the testes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.