Learn More
American Radiolabeled Chemicals Inc WIN 35428 14C 50UC
SDP
Catalog No. 50353047
Encompass
WIN 35,428-14C 50uCi in ethanol, 0.1 mCi/ml, 50-60 mCi/mmol, (B-CFT ), M.W. 277.3, Shipped in Dry Ice, Exclusive
Catalog No. 50-353-047 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 357850UC
Add to Cart
Add to Cart
Request a Quote
WIN 35,428-14C 50uCi in ethanol, 0.1 mCi/ml, 50-60 mCi/mmol, (B-CFT ), M.W. 277.3, Shipped in Dry Ice, Exclusive