Learn More
Cayman Chemical WIN 55 212-3mesylate 10mg
SDP
Catalog No. 502954957
Encompass_Preferred
A competitive neutral antagonist of CB2; blocks the stimulating action of CP 55,940 (pA2 = 6.1) and the inverse agonism of SR 144528 (pEC50 = 5.3); weakly antagonizes the melatonin MT1 and muscarinic M4 receptors
Catalog No. 50-295-4957 Supplier Cayman Chemical Supplier No. 1202210 MG
A competitive neutral antagonist of CB2; blocks the stimulating action of CP 55,940 (pA2 = 6.1) and the inverse agonism of SR 144528 (pEC50 = 5.3); weakly antagonizes the melatonin MT1 and muscarinic M4 receptors