Learn More
Cayman Chemical WIN 55 212 mesylate
SDP
Supplier:  Cayman Chemical 1073650 MG
Encompass_Preferred
A racemic mixture of (+)-WIN 55,212-2 and (−)-WIN 55,212-3; inhibits electrically induced contractions in mouse vas deferens preparations (IC50 = 0.006 µM) and reduces PG synthesis by 35% in mouse brain microsomes at 30 µM; decreases acetylcholine-induced writhing in mice (ED50 = 0.25 mg/kg); induces hypolocomotion, analgesia, hypothermia, and catalepsy in mice (ED50s = 1.09, 0.5, 3.01 and 6.8 mg/kg, respectively)
Catalog No. 50-288-7996