Learn More
American Radiolabeled Chemicals Inc WIN 55212-2 3H 250UC
SDP
Catalog No. 50901135
Encompass
WIN 55,212-2-5,7-naphthyl-3H 250uCi in ethanol, 1 mCi/ml, S.A. 40-60 Ci/mmol, M.W. 426, Shipped in Dry Ice
Catalog No. 50-901-135 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1806250UC
Add to Cart
Add to Cart
Request a Quote
WIN 55,212-2-5,7-naphthyl-3H 250uCi in ethanol, 1 mCi/ml, S.A. 40-60 Ci/mmol, M.W. 426, Shipped in Dry Ice