Learn More
American Radiolabeled Chemicals Inc WIN 55212-2 3H 25UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 180625UC
Encompass
WIN 55,212-2-5,7-naphthyl-3H 25uCi in ethanol, 1 mCi/ml, S.A. 40-60 Ci/mmol, M.W. 426, Shipped in Dry Ice
Catalog No. 50-901-134