Learn More
Grainger Winch Bar 38 5/8 L Steel Painted
Catalog No. 502766414
Encompass_Preferred
The ANCRA Winch Bar is a steel pry bar made in the USA. It has a hexagonal cross section and measures 36 inches long with a 1-1/8 inch diameter. The winch bar is made from drop-forged 8620 alloy steel and has a black oxide finish. It can provide up to 5,000 lbs of prying force. The winch bar weighs 9 lbs and has a limited lifetime warranty. It is designed for heavy duty prying applications.
Catalog No. 50-276-6414 Supplier Grainger Supplier No. 16V927
The ANCRA Winch Bar is a steel pry bar made in the USA. It has a hexagonal cross section and measures 36 inches long with a 1-1/8 inch diameter. The winch bar is made from drop-forged 8620 alloy steel and has a black oxide finish. It can provide up to 5,000 lbs of prying force. The winch bar weighs 9 lbs and has a limited lifetime warranty. It is designed for heavy duty prying applications.