Learn More
Cole-Parmer R. M. Young 5103 Wind Monitor Sensor, wind speed and direction
Catalog No. 50992501 Shop All Cole-Parmer Products
Encompass_Preferred
The Wind Monitor is a high resolution rugged wind sensor. Its simplicity and lightweight corrosion resistant construction make it well suited for a wide range of wind measuring applications. The wind speed sensor is a four blade helicoid propeller. Propeller rotation produces an AC sine wave voltage signal with frequency directly proportional to wind speed. Slip rings and brushes are not used resulting in improved reliability and lower cost. The wind direction sensor is a rugged yet lightweight vane with a sufficiently low aspect ratio to assure good fidelity in fluctuating wind conditions. Vane position is sensed by a precision conductive plastic potentiometer located in a sealed chamber.
Catalog No. 50-992-501 Supplier Cole-Parmer Supplier No. 9953618
The Wind Monitor is a high resolution rugged wind sensor. Its simplicity and lightweight corrosion resistant construction make it well suited for a wide range of wind measuring applications. The wind speed sensor is a four blade helicoid propeller. Propeller rotation produces an AC sine wave voltage signal with frequency directly proportional to wind speed. Slip rings and brushes are not used resulting in improved reliability and lower cost. The wind direction sensor is a rugged yet lightweight vane with a sufficiently low aspect ratio to assure good fidelity in fluctuating wind conditions. Vane position is sensed by a precision conductive plastic potentiometer located in a sealed chamber.