Learn More
Supertek™ Wind Power Demonstrator
GREENER_CHOICESCI_ED
Catalog No. S160130 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (L x W x H):
10.24 x 7.68 x 8.66 in.
Catalog No. Dimensions (L x W x H)
S160130 10.24 x 7.68 x 8.66 in.
1 options
This item is not returnable. View return policy
Catalog No. S160130 Supplier Supertek™ Supplier No. PH20450
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Supertek™ Wind Power Demonstrator can be used to demonstrate the generation of electricity by a wind-powered turbine.
  • Wind produced by the hand cranked fan, subsequently turns the second fan connected to a generator.
  • When the fan rotates, it produces electricity and lights up a small LED and a voltmeter.
  • Instructions included.
Specifications
Product Type Wind Power Generator
Dimensions (L x W x H) 10.24 x 7.68 x 8.66 in.