Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Wind Tunnel
SCI_ED

Catalog No. S38980 Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S38980 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S38980 Supplier GSC Supplier No. WNDTNL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSC Wind Tunnel allows students to investigate lift, Bernouli’s principle, and aerodynamics

  • Simply connect the tunnel to a power source (sold separately), turn the fan on, and place a source of smoke in front of the fan
  • Watch the smoke move at different rates on either side of the wing
  • Design allows the student to build their own unique wing design and test it for aerodynamics
  • Banana jacks allow for easy connection to a power supply

Specifications

Product Type Physical Science
Quantity 1
Length (English) 40 in.
Width (Metric) 20 cm
Weight (Metric) 1.67 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.