Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Windaus ketone | 55812-80-1 | 98.4% | C19H32O | 100 MG
SDP

Catalog No. 5000358200
Encompass_Preferred

Windaus ketone is identified as a VD building blocker. This compound features:

  • Molecular formula C19H32O
  • Molecular weight 276.46
  • Targeting VD/VDR pathway
  • Purity of 98.4%
  • Appearance as a colorless to light yellow oil
  • Storage in pure form at -20°C for 3 years or 4°C for 2 years
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-58200 Supplier Medchemexpress LLC Supplier No. HY52121100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Windaus ketone is identified as a VD building blocker. This compound features:

  • Molecular formula C19H32O
  • Molecular weight 276.46
  • Targeting VD/VDR pathway
  • Purity of 98.4%
  • Appearance as a colorless to light yellow oil
  • Storage in pure form at -20°C for 3 years or 4°C for 2 years
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.