Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Windaus ketone | 55812-80-1 | 98.4% | C19H32O | 25 MG
SDP

Catalog No. 5000358406
Encompass_Preferred

Windaus ketone is a VD building blocker identified for use as a laboratory chemical in the manufacture of substances. This product appears as a colorless to light yellow oil and is stable under recommended storage conditions.

  • Targeted to VD/VDR pathway
  • Soluble in DMSO (≥ 100 mg/mL)
  • Not classified as a hazardous substance or mixture
  • Purity of 98.4% (HPLC)

Catalog No. 50-003-58406 Supplier Medchemexpress LLC Supplier No. 25MGHY52121
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Windaus ketone is a VD building blocker identified for use as a laboratory chemical in the manufacture of substances. This product appears as a colorless to light yellow oil and is stable under recommended storage conditions.

  • Targeted to VD/VDR pathway
  • Soluble in DMSO (≥ 100 mg/mL)
  • Not classified as a hazardous substance or mixture
  • Purity of 98.4% (HPLC)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.