Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Windaus ketone | 55812-80-1 | C19H32O | 1 ML
SDP

Catalog No. 5000357788
Encompass_Preferred

Windaus ketone is a VD building blocker. This chemical is stable under recommended storage conditions and is for research use only. It should be kept in a cool, well-ventilated area, away from direct sunlight and sources of ignition.

  • Stable under recommended storage conditions
  • VD building blocker
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks, then store at -20°C for 1 month

Catalog No. 50-003-57788 Supplier Medchemexpress LLC Supplier No. HY5212110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Windaus ketone is a VD building blocker. This chemical is stable under recommended storage conditions and is for research use only. It should be kept in a cool, well-ventilated area, away from direct sunlight and sources of ignition.

  • Stable under recommended storage conditions
  • VD building blocker
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks, then store at -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.