Learn More
Medchemexpress LLC Windaus ketone | 55812-80-1 | C19H32O | 50 MG
SDP
Catalog No. 5000358331
Encompass_Preferred
Windaus ketone is identified as a VD building blocker, acting on the VD/VDR target within the Vitamin D Related/Nuclear Receptor pathway.
  • Acts as a VD building blocker
  • Targets VD/VDR
  • Functions within the Vitamin D Related/Nuclear Receptor pathway
  • Soluble in DMSO at concentrations of 100 mg/mL (361.72 mM) or greater
  • Pure form can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-58331 Supplier Medchemexpress LLC Supplier No. HY5212150MG
Windaus ketone is identified as a VD building blocker, acting on the VD/VDR target within the Vitamin D Related/Nuclear Receptor pathway.
  • Acts as a VD building blocker
  • Targets VD/VDR
  • Functions within the Vitamin D Related/Nuclear Receptor pathway
  • Soluble in DMSO at concentrations of 100 mg/mL (361.72 mM) or greater
  • Pure form can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month