Learn More
Medchemexpress LLC Windaus ketone | 55812-80-1 | MFCD22573278 | 98.4% | 276.46 g/mol | C19H32O | 10 MG
SDP
Catalog No. 5000298729
Encompass_Preferred
Windaus ketone is a vitamin D building block and VDR antagonist provided for research use. It is supplied as an oil or as a DMSO stock solution and is intended for use in synthetic and biochemical studies involving vitamin D pathways.
  • Research-grade building block for vitamin D synthesis and VDR antagonist studies.
  • High purity (≈98.4%) supporting reproducible experimental results.
  • Available as oil and as a 10 mM solution in DMSO for convenient handling.
  • Storage-stable under recommended conditions for long-term use.
  • Supplied in small-scale quantities suitable for bench-scale synthesis and assay work.
Catalog No. 50-002-98729 Supplier Medchemexpress LLC Supplier No. HY5212110MG
Windaus ketone is a vitamin D building block and VDR antagonist provided for research use. It is supplied as an oil or as a DMSO stock solution and is intended for use in synthetic and biochemical studies involving vitamin D pathways.
  • Research-grade building block for vitamin D synthesis and VDR antagonist studies.
  • High purity (≈98.4%) supporting reproducible experimental results.
  • Available as oil and as a 10 mM solution in DMSO for convenient handling.
  • Storage-stable under recommended conditions for long-term use.
  • Supplied in small-scale quantities suitable for bench-scale synthesis and assay work.