Learn More
MSC LG Window Air Conditioner with Electric Heat, 7600 BTU, 115V, 6.3A, 1 ea
Catalog No. 5000507852
Encompass
This window-mounted air conditioning unit provides both cooling and heating capabilities for residential spaces. Engineered for efficiency and ease of use, it covers an area of up to 330 sq. ft. and features an intuitive control system with an included remote. The unit is designed for simple installation and reliable operation, offering versatile climate control in a compact white chassis.
  • Provides 7,600 BTU of cooling and 3,850 BTU of heating capacity
  • Features 2 cooling speeds, 2 heating speeds, and 2 fan speeds
  • Includes a 24-hour timer and 4-way air deflection
  • Equipped with an energy saver mode and auto restart function
  • Includes a removable, washable filter with a check filter alert
  • Operates at 115V and 6.3 A with an EER rating of 11.3
Catalog No. 50-005-07852 Supplier MSC Supplier No. 67409789
This window-mounted air conditioning unit provides both cooling and heating capabilities for residential spaces. Engineered for efficiency and ease of use, it covers an area of up to 330 sq. ft. and features an intuitive control system with an included remote. The unit is designed for simple installation and reliable operation, offering versatile climate control in a compact white chassis.
  • Provides 7,600 BTU of cooling and 3,850 BTU of heating capacity
  • Features 2 cooling speeds, 2 heating speeds, and 2 fan speeds
  • Includes a 24-hour timer and 4-way air deflection
  • Equipped with an energy saver mode and auto restart function
  • Includes a removable, washable filter with a check filter alert
  • Operates at 115V and 6.3 A with an EER rating of 11.3