Learn More
Chemplex Industries WINDOW FRAME 0.1 MIL 3.0IN DIA
Catalog No. 50249349
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 2.5 u (0.1 mil), 3.0 Dia., In.
Catalog No. 50-249-349 Supplier Chemplex Industries Supplier No. 3012
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 2.5 u (0.1 mil), 3.0 Dia., In.