Learn More
Chemplex Industries Etnom 3.0 μm (0.12 mil); 3.0"; SpectroMembrane Thin-Film Sample Supports
Catalog No. 50249365
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Etnom(TM), 3.0 u (0.12 mil), 3.0 Dia., In.
Catalog No. 50-249-365 Supplier Chemplex Industries Supplier No. 3026
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Etnom(TM), 3.0 u (0.12 mil), 3.0 Dia., In.