Learn More
Chemplex Industries SpectroMembrane Mylar Thin Film Sample Supports" 3.6 μm (0.14 mil); 3.0"
Catalog No. 50249351
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 3.6 u (0.14 mil), 3.0 Dia., In.
Catalog No. 50-249-351 Supplier Chemplex Industries Supplier No. 3014
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 3.6 u (0.14 mil), 3.0 Dia., In.