Learn More
Chemplex Industries WINDOW FRAME 0.24 MIL 3.0IN DI
Catalog No. 50249353
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 6.0 u (0.24 mil), 3.0 Dia., In.
Catalog No. 50-249-353 Supplier Chemplex Industries Supplier No. 3016
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 6.0 u (0.24 mil), 3.0 Dia., In.