Learn More
Chemplex Industries WINDOW FRAME 0.50 MIL 2.5IN DI
Catalog No. 50249362
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Polypropylene, 12.0u (0.50 mil), 2.5 Dia., In.
Catalog No. 50-249-362 Supplier Chemplex Industries Supplier No. 3023
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Polypropylene, 12.0u (0.50 mil), 2.5 Dia., In.