Learn More
Beckman Coulter Microfuge 18 Centrifuge Tubes, 1.5mL, Natural, Bag of 500
Supplier:  Beckman Coulter 362329
Encompass
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 x g. The rotor accommodates 18 x 1.5/2.0mL microcentrifuge tubes. It features a compact footprint, digital display, 10 acceleration and braking rates, and a built-in 30 minute timer with hold function. This microcentrifuge is ideal for quick spin downs in life science labs with limited benchtop space.
Catalog No. NC9439139