Learn More
Microsolv Technology Corporation Window Maker Us Model 110/120V
Catalog No. 5001452758
Encompass_Preferred
Window Maker, Polyimide Coating Remover, 110/120v for 2mm Detection Windows for CE. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52758 Supplier Microsolv Technology Corporation Supplier No. 07200S
Window Maker, Polyimide Coating Remover, 110/120v for 2mm Detection Windows for CE. MicroSolv Brand. 1/EA.