Learn More
MSC Bare Ground Windshield Deicing Spray, Potassium Formate, 1 ea
Catalog No. 502869235
Encompass
This de-icing spray is an easy-to-use solution that utilizes a non-flammable nitrogen propellant and potassium formate deicing liquid, similar to what is used at airports. This environmentally friendly, biodegradable, and non-toxic solution quickly melts ice and frost from windshields and locks, saving time and energy.
  • Aerosol can, 16 oz
  • Non-flammable nitrogen propellant
  • Uses potassium formate deicing liquid
  • Environmentally friendly and biodegradable
  • Gentle on all surfaces
  • Non-toxic
  • Quickly melts ice and frost
  • Suitable for windshields, locks, and more
  • Helps prevent ice formation
  • Improves visibility
  • Saves time and energy by reducing defrosting needs
Catalog No. 50-286-9235 Supplier MSC Supplier No. 55598742
This de-icing spray is an easy-to-use solution that utilizes a non-flammable nitrogen propellant and potassium formate deicing liquid, similar to what is used at airports. This environmentally friendly, biodegradable, and non-toxic solution quickly melts ice and frost from windshields and locks, saving time and energy.
  • Aerosol can, 16 oz
  • Non-flammable nitrogen propellant
  • Uses potassium formate deicing liquid
  • Environmentally friendly and biodegradable
  • Gentle on all surfaces
  • Non-toxic
  • Quickly melts ice and frost
  • Suitable for windshields, locks, and more
  • Helps prevent ice formation
  • Improves visibility
  • Saves time and energy by reducing defrosting needs