Learn More
Sentry Air Systems Model 200 Winged Sentry with Lid
SDP
Catalog No. NC2107092
Encompass
The Winged Sentry with Lid provides a benchtop fume control solution for chemical fumes, dust, and other lab processes. The adjustable wing flaps allow the operator to easily move to suit their application. When used with the acrylic lid, the Winged Sentry creates a hood-like enclosure protecting the operator from fumes and particulate. The wing flaps, lid, and powerful fan work together to direct fumes into the filtration system away from the operator’s breathing zone. After filtration, the system releases the air back into the surrounding room creating a recirculating air flow pattern. Approximate Airflow: up to 80 CFM. Filters Available: HEPA Filter [up to 99.97% efficiency on particles 0.3 microns and larger], ASHRAE Filter [up to 95% efficiency on particles 0.5 microns and larger], ULPA Filter [up to 99.995% efficiency on particles 0.13 microns and larger], Activated Carbon, and specialty-blended filtration [i.e. acid gas, mercury, aldehyde, and ammonia].
This item is not returnable. View return policy
Catalog No. NC2107092 Supplier Sentry Air Systems Supplier No. SS200WSL
The Winged Sentry with Lid provides a benchtop fume control solution for chemical fumes, dust, and other lab processes. The adjustable wing flaps allow the operator to easily move to suit their application. When used with the acrylic lid, the Winged Sentry creates a hood-like enclosure protecting the operator from fumes and particulate. The wing flaps, lid, and powerful fan work together to direct fumes into the filtration system away from the operator’s breathing zone. After filtration, the system releases the air back into the surrounding room creating a recirculating air flow pattern. Approximate Airflow: up to 80 CFM. Filters Available: HEPA Filter [up to 99.97% efficiency on particles 0.3 microns and larger], ASHRAE Filter [up to 95% efficiency on particles 0.5 microns and larger], ULPA Filter [up to 99.995% efficiency on particles 0.13 microns and larger], Activated Carbon, and specialty-blended filtration [i.e. acid gas, mercury, aldehyde, and ammonia].