Learn More
Electron Microscopy Sciences Winlab Slide Rack LS-20, 20 Slide Capacity
SDP
Catalog No. 50931054
Encompass_Preferred
WINLAB Racks are made from Poly-ceramic that maintains its structural integrity to 200 degree C in any solvent while being microwaved. Designed for histology and cytology staining and coverslipping. They last a lot longer than any other materials and save time and money. Fitted to a Sakura Staining Dish, either Green or White and also fits most other staining dishes.
Capacity: 20 slides
Catalog No. 50-931-054 Supplier Electron Microscopy Sciences Supplier No. 7143020
WINLAB Racks are made from Poly-ceramic that maintains its structural integrity to 200 degree C in any solvent while being microwaved. Designed for histology and cytology staining and coverslipping. They last a lot longer than any other materials and save time and money. Fitted to a Sakura Staining Dish, either Green or White and also fits most other staining dishes.
Capacity: 20 slides